Assay Details
| Gene Symbol: M17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_129691.3 | NP_181659.2 | 1 - 2 | 155 | 64 | |
| NM_201927.3 | NP_973656.1 | 1 - 2 | 182 | 64 | ||
| GenBank mRNA | AY128274.1 | - | 1 - 2 | 121 | 64 | |
More Information
Set Membership: |
|

M17
Genomic Map