Assay Details
| Gene Symbol: AT2G41480 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_129711.4 | NP_181679.4 | 1 - 2 | 254 | 81 | |
| GenBank mRNA | AY139994.1 | - | 1 - 2 | 256 | 81 | |
| BT008727.1 | - | 1 - 2 | 219 | 81 | ||
More Information
Set Membership: |
|

AT2G41480
Genomic Map