Assay Details
| Gene Symbol: GPT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_129712.3 | NP_565950.1 | 8 - 9 | 1263 | 82 | |
| GenBank mRNA | AF412104.1 | - | 8 - 9 | 1140 | 82 | |
| AY090264.1 | - | 8 - 9 | 1073 | 82 | ||
| D88036.1 | - | 8 - 9 | 1130 | 82 | ||
More Information
Set Membership: |
|

GPT
Genomic Map