Assay Details
| Gene Symbol: ACA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_129719.4 | NP_181687.1 | 1 - 2 | 486 | 81 | |
| GenBank mRNA | AF200739.1 | - | 1 - 2 | 190 | 81 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ACA4| Gene Symbol: ACA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_129719.4 | NP_181687.1 | 1 - 2 | 486 | 81 | |
| GenBank mRNA | AF200739.1 | - | 1 - 2 | 190 | 81 | |
Set Membership: |
|