Assay Details
| Gene Symbol: ACA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001336925.1 | NP_001323669.1 | 3 - 4 | 3559 | 83 | |
| NM_001336926.1 | NP_001323668.1 | 2 - 3 | 2192 | 83 | ||
| NM_001336927.1 | NP_001323670.1 | 2 - 3 | 3325 | 83 | ||
| NM_129719.4 | NP_181687.1 | 3 - 4 | 2470 | 83 | ||
| GenBank mRNA | AF200739.1 | - | 3 - 4 | 2174 | 83 | |
More Information
Set Membership: |
|

ACA4
Genomic Map