Assay Details
| Gene Symbol: GK-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001336938.1 | NP_001323627.1 | 4 - 5 | 536 | 113 | |
| NM_001336939.1 | NP_001323629.1 | 4 - 5 | 535 | 113 | ||
| NM_001336940.1 | NP_001323628.1 | 4 - 5 | 535 | 113 | ||
| NM_001336941.1 | NP_001323630.1 | 4 - 5 | 518 | 113 | ||
| NM_001336942.1 | NP_001323631.1 | 4 - 5 | 492 | 113 | ||
| NM_001336943.1 | NP_001323632.1 | 4 - 5 | 479 | 113 | ||
| NM_001336944.1 | NP_001323633.1 | 4 - 5 | 478 | 113 | ||
| NM_001336945.1 | NP_001323634.1 | 4 - 5 | 472 | 113 | ||
| NM_129752.5 | NP_565961.1 | 4 - 5 | 536 | 113 | ||
| GenBank mRNA | BT028912.1 | - | 2 - 3 | 199 | 113 | |
More Information
Set Membership: |
|

GK-1
Genomic Map