Assay Details
| Gene Symbol: GK-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001336938.1 | NP_001323627.1 | 5 - 6 | 658 | 59 | |
| NM_001336939.1 | NP_001323629.1 | 5 - 6 | 657 | 59 | ||
| NM_001336940.1 | NP_001323628.1 | 5 - 6 | 657 | 59 | ||
| NM_001336941.1 | NP_001323630.1 | 5 - 6 | 640 | 59 | ||
| NM_001336942.1 | NP_001323631.1 | 5 - 6 | 614 | 59 | ||
| NM_001336943.1 | NP_001323632.1 | 5 - 6 | 601 | 59 | ||
| NM_001336944.1 | NP_001323633.1 | 5 - 6 | 600 | 59 | ||
| NM_001336945.1 | NP_001323634.1 | 5 - 6 | 594 | 59 | ||
| NM_129752.5 | NP_565961.1 | 5 - 6 | 658 | 59 | ||
| GenBank mRNA | BT028912.1 | - | 3 - 4 | 321 | 59 | |
More Information
Set Membership: |
|

GK-1
Genomic Map