Assay Details
| Gene Symbol: POLD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_129775.3 | NP_181742.2 | 4 - 5 | 491 | 65 | |
| NM_201935.3 | NP_973664.1 | 4 - 5 | 693 | 65 | ||
| GenBank mRNA | AK221655.1 | - | 4 - 5 | 488 | 65 | |
| AY136418.1 | - | 4 - 5 | 488 | 65 | ||
| BT008753.1 | - | 4 - 5 | 421 | 65 | ||
More Information
Set Membership: |
|

POLD2
Genomic Map