Assay Details
| Gene Symbol: MBF1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_129829.4 | NP_565981.1 | 2 - 3 | 366 | 96 | |
| GenBank mRNA | AF370280.1 | - | 2 - 3 | 290 | 96 | |
| AY063014.1 | - | 2 - 3 | 226 | 96 | ||
| AY088140.1 | - | 2 - 3 | 295 | 96 | ||
More Information
Set Membership: |
|

MBF1A
Genomic Map