Assay Details
| Gene Symbol: AT2G42700 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202805.1 | NP_001189734.1 | 1 - 2 | 233 | 84 | |
| NM_129831.5 | NP_181798.2 | 1 - 2 | 377 | 84 | ||
More Information
Set Membership: |
|

AT2G42700
Genomic Map