Assay Details
| Gene Symbol: AT2G42700 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202805.1 | NP_001189734.1 | 4 - 5 | 981 | 136 | |
| NM_001336992.1 | NP_001323776.1 | 3 - 4 | 795 | 136 | ||
| NM_129831.5 | NP_181798.2 | 4 - 5 | 1125 | 136 | ||
More Information
Set Membership: |
|

AT2G42700
Genomic Map