Assay Details
| Gene Symbol: AT-P4H-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001337014.1 | NP_001324186.1 | 11 - 12 | 964 | 70 | |
| NM_129869.5 | NP_181836.1 | 11 - 12 | 1052 | 70 | ||
| GenBank mRNA | AK117688.1 | - | 11 - 12 | 829 | 70 | |
| BT006098.1 | - | 11 - 12 | 745 | 70 | ||
More Information
Set Membership: |
|

AT-P4H-1
Genomic Map