Assay Details
| Gene Symbol: AT-P4H-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001337014.1 | NP_001324186.1 | 4 - 5 | 441 | 111 | |
| NM_129869.5 | NP_181836.1 | 4 - 5 | 534 | 111 | ||
| GenBank mRNA | AK117688.1 | - | 4 - 5 | 311 | 111 | |
| BT006098.1 | - | 4 - 5 | 227 | 111 | ||
More Information
Set Membership: |
|

AT-P4H-1
Genomic Map