Assay Details
| Gene Symbol: AT-P4H-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001337014.1 | NP_001324186.1 | 7 - 8 | 656 | 91 | |
| NM_129869.5 | NP_181836.1 | 7 - 8 | 749 | 91 | ||
| GenBank mRNA | AK117688.1 | - | 7 - 8 | 526 | 91 | |
| BT006098.1 | - | 7 - 8 | 442 | 91 | ||
More Information
Set Membership: |
|

AT-P4H-1
Genomic Map