Assay Details
| Gene Symbol: AT2G43420 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_129903.3 | NP_565998.1 | 11 - 12 | 1776 | 66 | |
| GenBank mRNA | AK117930.1 | - | 11 - 12 | 1681 | 66 | |
| AY045841.1 | - | 11 - 12 | 1647 | 66 | ||
More Information
Set Membership: |
|

AT2G43420
Genomic Map