Assay Details
| Gene Symbol: GLX2-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001337029.1 | NP_001323646.1 | 3 - 4 | 545 | 120 | |
| NM_129904.3 | NP_565999.1 | 3 - 4 | 721 | 120 | ||
| NM_201950.3 | NP_973679.1 | 2 - 3 | 1101 | 120 | ||
| GenBank mRNA | AY063806.1 | - | 3 - 4 | 606 | 120 | |
| AY091278.1 | - | 3 - 4 | 451 | 120 | ||
More Information
Set Membership: |
|

GLX2-1
Genomic Map