Assay Details
| Gene Symbol: GLX2-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001337029.1 | NP_001323646.1 | 5 - 6 | 773 | 74 | |
| NM_129904.3 | NP_565999.1 | 5 - 6 | 949 | 74 | ||
| NM_201950.3 | NP_973679.1 | 4 - 5 | 1329 | 74 | ||
| GenBank mRNA | AY063806.1 | - | 5 - 6 | 834 | 74 | |
| AY091278.1 | - | 5 - 6 | 679 | 74 | ||
More Information
Set Membership: |
|

GLX2-1
Genomic Map