Assay Details
| Gene Symbol: GLX2-1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001337029.1 | NP_001323646.1 | 7 - 8 | 946 | 86 | |
| NM_129904.3 | NP_565999.1 | 7 - 8 | 1122 | 86 | ||
| NM_201950.3 | NP_973679.1 | 6 - 7 | 1502 | 86 | ||
| GenBank mRNA | AY063806.1 | - | 7 - 8 | 1007 | 86 | |
| AY091278.1 | - | 7 - 8 | 852 | 86 | ||
More Information
Set Membership: |
|

GLX2-1
Genomic Map