Assay Details
| Gene Symbol: AT2G43500 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161101.2 | NP_001154573.1 | 3 - 4 | 1197 | 126 | |
| NM_001337037.1 | NP_001318414.1 | 3 - 4 | 1367 | 126 | ||
| NM_001337038.1 | NP_001325296.1 | 1 - 2 | 1611 | 126 | ||
| NM_001337039.1 | NP_001325295.1 | 2 - 3 | 1517 | 126 | ||
| NM_001337040.1 | NP_001325294.1 | 2 - 3 | 1461 | 126 | ||
| NM_001337043.1 | NP_001325291.1 | 3 - 4 | 1274 | 126 | ||
| NM_001337044.1 | NP_001325290.1 | 3 - 4 | 1242 | 126 | ||
| NM_001337045.1 | NP_001325289.1 | 3 - 4 | 1196 | 126 | ||
| NM_001337046.1 | NP_001325288.1 | 3 - 4 | 1268 | 126 | ||
More Information
Set Membership: |
|

AT2G43500
Genomic Map