Assay Details
| Gene Symbol: AT2G43500 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161101.2 | NP_001154573.1 | 4 - 5 | 1229 | 62 | |
| NM_001337037.1 | NP_001318414.1 | 4 - 5 | 1399 | 62 | ||
| NM_001337038.1 | NP_001325296.1 | 2 - 3 | 1643 | 62 | ||
| NM_001337039.1 | NP_001325295.1 | 3 - 4 | 1549 | 62 | ||
| NM_001337040.1 | NP_001325294.1 | 3 - 4 | 1493 | 62 | ||
| NM_001337041.1 | NP_001325293.1 | 5 - 6 | 1349 | 62 | ||
| NM_001337042.1 | NP_001325292.1 | 5 - 6 | 1345 | 62 | ||
| NM_001337043.1 | NP_001325291.1 | 4 - 5 | 1306 | 62 | ||
| NM_001337044.1 | NP_001325290.1 | 4 - 5 | 1274 | 62 | ||
| NM_001337045.1 | NP_001325289.1 | 4 - 5 | 1228 | 62 | ||
| NM_001337046.1 | NP_001325288.1 | 4 - 5 | 1300 | 62 | ||
More Information
Set Membership: |
|

AT2G43500
Genomic Map