Assay Details
| Gene Symbol: AT2G43500 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161101.2 | NP_001154573.1 | 6 - 7 | 2119 | 74 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT2G43500| Gene Symbol: AT2G43500 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161101.2 | NP_001154573.1 | 6 - 7 | 2119 | 74 | |
Set Membership: |
|