Assay Details
| Gene Symbol: AT2G43500 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161101.2 | NP_001154573.1 | 7 - 8 | 2485 | 99 | |
| NM_001337037.1 | NP_001318414.1 | 7 - 8 | 2616 | 99 | ||
| NM_001337038.1 | NP_001325296.1 | 5 - 6 | 2860 | 99 | ||
| NM_001337039.1 | NP_001325295.1 | 6 - 7 | 2766 | 99 | ||
| NM_001337040.1 | NP_001325294.1 | 6 - 7 | 2710 | 99 | ||
| NM_001337041.1 | NP_001325293.1 | 8 - 9 | 2566 | 99 | ||
| NM_001337042.1 | NP_001325292.1 | 8 - 9 | 2562 | 99 | ||
| NM_001337043.1 | NP_001325291.1 | 7 - 8 | 2523 | 99 | ||
| NM_001337044.1 | NP_001325290.1 | 7 - 8 | 2491 | 99 | ||
| NM_001337045.1 | NP_001325289.1 | 7 - 8 | 2445 | 99 | ||
| NM_001337046.1 | NP_001325288.1 | 7 - 8 | 2517 | 99 | ||
More Information
Set Membership: |
|

AT2G43500
Genomic Map