Assay Details
| Gene Symbol: MPK6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_129941.4 | NP_181907.1 | 1 - 2 | 465 | 125 | |
| GenBank mRNA | AY120737.1 | - | 1 - 2 | 348 | 125 | |
| BT008855.1 | - | 1 - 2 | 234 | 125 | ||
| D21842.1 | - | 1 - 2 | 306 | 125 | ||
More Information
Set Membership: |
|

MPK6
Genomic Map