Assay Details
| Gene Symbol: MPK6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_129941.4 | NP_181907.1 | 4 - 5 | 1068 | 80 | |
| GenBank mRNA | AY120737.1 | - | 4 - 5 | 951 | 80 | |
| BT008855.1 | - | 4 - 5 | 837 | 80 | ||
| D21842.1 | - | 4 - 5 | 909 | 80 | ||
More Information
Set Membership: |
|

MPK6
Genomic Map