Assay Details
| Gene Symbol: LSM6B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084583.2 | NP_001078052.1 | 3 - 4 | 425 | 62 | |
| NM_001337061.1 | NP_001318419.1 | 3 - 4 | 510 | 62 | ||
| NM_129943.3 | NP_181909.1 | 4 - 5 | 371 | 62 | ||
| GenBank mRNA | AK227857.1 | - | 3 - 4 | 311 | 62 | |
| AK317095.1 | - | 4 - 5 | 405 | 62 | ||
| BT008305.1 | - | 2 - 3 | 221 | 62 | ||
More Information
Set Membership: |
|

LSM6B
Genomic Map