Assay Details
| Gene Symbol: UGT74F2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_129944.3 | NP_181910.1 | 1 - 2 | 825 | 128 | |
| GenBank mRNA | AY062483.1 | - | 1 - 2 | 667 | 128 | |
| AY087340.1 | - | 1 - 2 | 676 | 128 | ||
| BT010327.1 | - | 1 - 2 | 624 | 128 | ||
| DQ407524.1 | - | 1 - 2 | 624 | 128 | ||
| KJ138855.1 | - | 1 - 2 | 624 | 128 | ||
More Information
Set Membership: |
|

UGT74F2
Genomic Map