Assay Details
| Gene Symbol: MTHFR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_129979.3 | NP_566011.1 | 3 - 4 | 838 | 90 | |
| GenBank mRNA | AF181967.1 | - | 3 - 4 | 726 | 90 | |
| AY050434.1 | - | 3 - 4 | 766 | 90 | ||
More Information
Set Membership: |
|

MTHFR2
Genomic Map