Assay Details
| Gene Symbol: MAP2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001337079.1 | NP_001323713.1 | 1 - 2 | 333 | 136 | |
| NM_129981.5 | NP_566013.1 | 1 - 2 | 332 | 136 | ||
| GenBank mRNA | AK228284.1 | - | 1 - 2 | 235 | 136 | |
| BT006476.1 | - | 1 - 2 | 177 | 136 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001337079.1 transcripts.
Set Membership: |
|

MAP2A
Genomic Map