Assay Details
| Gene Symbol: MAP2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_129981.5 | NP_566013.1 | 3 - 4 | 476 | 97 | |
| GenBank mRNA | AK228284.1 | - | 3 - 4 | 379 | 97 | |
| BT006476.1 | - | 3 - 4 | 321 | 97 | ||
More Information
Set Membership: |
|

MAP2A
Genomic Map