Assay Details
| Gene Symbol: MAP2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001337079.1 | NP_001323713.1 | 4 - 5 | 880 | 68 | |
| NM_129981.5 | NP_566013.1 | 5 - 6 | 812 | 68 | ||
| GenBank mRNA | AK228284.1 | - | 5 - 6 | 715 | 68 | |
| BT006476.1 | - | 5 - 6 | 657 | 68 | ||
More Information
Set Membership: |
|

MAP2A
Genomic Map