Assay Details
| Gene Symbol: EMB2761 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001335263.1 | NP_001318198.1 | 7 - 8 | 1851 | 73 | |
| NM_147245.2 | NP_671778.1 | 7 - 8 | 1863 | 73 | ||
More Information
Set Membership: |
|

EMB2761
Genomic Map