Assay Details
| Gene Symbol: AT5G51105 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_148116.4 | NP_680421.1 | 1 | 243 | 77 | |
| Gene Symbol: AT5G51110 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_124490.5 | NP_199924.1 | 3 | 1271 | 77 | |
More Information
Set Membership: |
|

AT5G51105
Genomic Map