Assay Details
| Gene Symbol: MYB82 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_148121.3 | NP_680426.1 | 2 - 3 | 399 | 150 | |
| GenBank mRNA | AY519637.1 | - | 2 - 3 | 261 | 150 | |
More Information
Set Membership: |
|

MYB82
Genomic Map