Assay Details
| Gene Symbol: AT4G12545 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_179035.1 | NP_849366.1 | 1 | 582 | 153 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT4G12545| Gene Symbol: AT4G12545 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_179035.1 | NP_849366.1 | 1 | 582 | 153 | |
Set Membership: |
|