Assay Details
| Gene Symbol: SCRL5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_202329.3 | NP_974058.1 | 1 - 2 | 150 | 99 | |
| GenBank mRNA | AY735544.1 | - | 1 - 2 | 138 | 99 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SCRL5| Gene Symbol: SCRL5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_202329.3 | NP_974058.1 | 1 - 2 | 150 | 99 | |
| GenBank mRNA | AY735544.1 | - | 1 - 2 | 138 | 99 | |
Set Membership: |
|