Assay Details
| Gene Symbol: La2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_202456.2 | NP_974185.1 | 9 - 10 | 1257 | 104 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

La2| Gene Symbol: La2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_202456.2 | NP_974185.1 | 9 - 10 | 1257 | 104 | |
Set Membership: |
|