Assay Details
| Gene Symbol: AT4G37235 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_202971.2 | NP_974700.1 | 2 - 3 | 581 | 116 | |
| GenBank mRNA | BT015421.1 | - | 2 - 3 | 417 | 116 | |
More Information
Set Membership: |
|

AT4G37235
Genomic Map