Assay Details
| Gene Symbol: IDH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_127267.3 | NP_179304.1 | 2 - 3 | 894 | 145 | |
| NM_179632.2 | NP_849963.1 | 2 - 3 | 882 | 145 | ||
| GenBank mRNA | AK228337.1 | - | 2 - 3 | 842 | 145 | |
More Information
Set Membership: |
|

IDH2
Genomic Map