Assay Details
| Gene Symbol: SDR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084482.3 | NP_001077951.1 | 4 - 5 | 596 | 162 | |
| NM_127980.4 | NP_179996.1 | 4 - 5 | 667 | 162 | ||
More Information
Set Membership: |
|

SDR2
Genomic Map