Assay Details
| Gene Symbol: CAM5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_128261.5 | NP_180271.1 | 1 - 2 | 174 | 117 | |
| NM_179766.4 | NP_850097.1 | 1 - 2 | 174 | 117 | ||
| GenBank mRNA | AK175278.1 | - | 1 - 2 | 134 | 117 | |
| AK176855.1 | - | 1 - 2 | 132 | 117 | ||
| BT002351.1 | - | 1 - 2 | 72 | 117 | ||
| M38380.1 | - | 789 | 117 | |||
More Information
Set Membership: |
|

CAM5
Genomic Map