Assay Details
| Gene Symbol: MT1C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_100634.2 | NP_172240.1 | 1 - 2 | 211 | 166 | |
| GenBank mRNA | AF375400.1 | - | 1 - 2 | 116 | 166 | |
| AY085074.1 | - | 1 - 2 | 116 | 166 | ||
More Information
Set Membership: |
|

MT1C
Genomic Map