Assay Details
| Gene Symbol: ECA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001331716.1 | NP_001321027.1 | 3 - 4 | 1847 | 120 | |
| NM_100640.4 | NP_172246.3 | 4 - 5 | 1765 | 120 | ||
More Information
Set Membership: |
|

ECA4
Genomic Map