Assay Details
| Gene Symbol: ECA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_100655.4 | NP_172259.1 | 4 - 5 | 1818 | 121 | |
| GenBank mRNA | U93845.1 | - | 3 - 4 | 1678 | 121 | |
| U96455.1 | - | 3 - 4 | 1674 | 121 | ||
More Information
Set Membership: |
|

ECA1
Genomic Map