Assay Details
| Gene Symbol: ECA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_100655.4 | NP_172259.1 | 8 - 9 | 2814 | 139 | |
| GenBank mRNA | U93845.1 | - | 7 - 8 | 2674 | 139 | |
| U96455.1 | - | 7 - 8 | 2670 | 139 | ||
More Information
Set Membership: |
|

ECA1
Genomic Map