Assay Details
| Gene Symbol: AT1G17010 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101561.3 | NP_173144.1 | 3 - 4 | 938 | 166 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT1G17010| Gene Symbol: AT1G17010 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_101561.3 | NP_173144.1 | 3 - 4 | 938 | 166 | |
Set Membership: |
|