Assay Details
| Gene Symbol: MYB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001332555.1 | NP_001322405.1 | 1 - 2 | 202 | 122 | |
| NM_102111.4 | NP_564176.2 | 2 - 3 | 484 | 122 | ||
| GenBank mRNA | AY072543.1 | - | 2 - 3 | 352 | 122 | |
| BT000841.1 | - | 2 - 3 | 259 | 122 | ||
More Information
Set Membership: |
|

MYB3
Genomic Map