Assay Details
| Gene Symbol: ASB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198163.1 | NP_001185092.1 | 1 - 2 | 181 | 151 | |
| NM_102331.3 | NP_173893.1 | 1 - 2 | 241 | 151 | ||
| GenBank mRNA | AK226500.1 | - | 1 - 2 | 137 | 151 | |
| AY099834.1 | - | 1 - 2 | 132 | 151 | ||
| BT006303.1 | - | 1 - 2 | 110 | 151 | ||
| L22585.1 | - | 1 - 2 | 126 | 151 | ||
More Information
Set Membership: |
|

ASB1
Genomic Map