Assay Details
| Gene Symbol: ACD6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_117519.4 | NP_567430.1 | 3 - 4 | 1202 | 144 | |
| NM_179050.2 | NP_849381.1 | 2 - 3 | 838 | 144 | ||
| NM_179051.1 | NP_849382.1 | 3 - 4 | 902 | 144 | ||
| GenBank mRNA | AK176809.1 | - | 3 - 4 | 915 | 144 | |
| AK316751.1 | - | 3 - 4 | 903 | 144 | ||
| AY094416.1 | - | 3 - 4 | 917 | 144 | ||
| AY344843.1 | - | 3 - 4 | 916 | 144 | ||
| BT002281.1 | - | 2 - 3 | 822 | 144 | ||
More Information
Set Membership: |
|

ACD6
Genomic Map