Assay Details
| Gene Symbol: UBC9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_118934.3 | NP_567791.1 | 1 - 2 | 367 | 146 | |
| NM_179131.3 | NP_849462.1 | 2 - 3 | 265 | 146 | ||
| GenBank mRNA | DQ027023.1 | - | 1 - 2 | 160 | 146 | |
| L00639.1 | - | 1 - 2 | 120 | 146 | ||
| X72626.1 | - | 104 | 146 | |||
| Z14990.1 | - | 120 | 146 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_179131.3, X72626.1, Z14990.1 transcripts.
Set Membership: |
|

UBC9
Genomic Map