Assay Details
| Gene Symbol: RPL16A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_180045.4 | NP_850376.2 | 3 - 4 | 250 | 131 | |
| GenBank mRNA | AK228536.1 | - | 3 - 4 | 183 | 131 | |
More Information
Set Membership: |
|

RPL16A
Genomic Map